siRNA Details
| Gene Symbol: PLXNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105543.1 | NP_001099013.1 | 2 | 813 | ||
| NM_020911.1 | NP_065962.1 | 2 | 813 | |||
| NM_181775.3 | NP_861440.2 | 3 | 961 | |||
| XM_005250686.4 | XP_005250743.1 | 2 | 1211 | |||
| XM_006716171.3 | XP_006716234.1 | 3 | 1304 | |||
| XM_011516676.2 | XP_011514978.1 | 2 | 719 | |||
| XM_017012779.1 | XP_016868268.1 | 2 | 719 | |||
| GenBank mRNA | AK123428.1 | - | 2 | 719 | ||
| BC028744.2 | - | 3 | 961 | |||

PLXNA4
Genomic Map