siRNA Details
| Gene Symbol: FER1L6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103547.1 | 4 | 946 | |||
| GenBank mRNA | AK057332.1 | - | 4 | 946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FER1L6-AS2| Gene Symbol: FER1L6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103547.1 | 4 | 946 | |||
| GenBank mRNA | AK057332.1 | - | 4 | 946 | ||