siRNA Details
Gene Symbol: KIAA1456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094299.1 | - | 4 | 1485 | ||
BC035082.1 | - | 3 | 1915 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094299.1 | - | 4 | 1485 | ||
BC035082.1 | - | 3 | 1915 |