siRNA Details
| Gene Symbol: FAM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163991.1 | NP_001157463.1 | 4 | 1187 | ||
| NM_182761.3 | NP_877438.2 | 5 | 1328 | |||
| GenBank mRNA | AF427126.1 | - | 5 | 1129 | ||
| AY692447.1 | - | 4 | 1187 | |||
| BC040622.1 | - | 5 | 1289 | |||

FAM170A
Genomic Map