siRNA Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182974.2 | NP_892019.2 | 4 | 477 | ||
| XM_011518633.1 | XP_011516935.1 | 6 | 693 | |||
| XM_011518634.1 | XP_011516936.1 | 5 | 632 | |||
| XM_011518635.1 | XP_011516937.1 | 6 | 598 | |||
| XM_011518636.1 | XP_011516938.1 | 5 | 537 | |||
| XM_011518637.1 | XP_011516939.1 | 6 | 693 | |||
| XM_011518638.2 | XP_011516940.1 | 2 | 202 | |||
| GenBank mRNA | AY336054.1 | - | 4 | 477 | ||

GLT6D1
Genomic Map