siRNA Details
| Gene Symbol: ARSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198150.2 | NP_937793.1 | 2 | 362 | ||
| XM_005271904.3 | XP_005271961.1 | 2 | 375 | |||
| GenBank mRNA | AK303855.1 | - | 2 | 414 | ||
| AK316253.1 | - | 2 | 382 | |||
| AL832711.1 | - | 1 | 45 | |||
| AY358596.1 | - | 2 | 322 | |||
| AY875939.1 | - | 2 | 204 | |||
| BC036047.1 | - | 2 | 224 | |||
| BC050392.1 | - | 2 | 321 | |||
| BC130329.1 | - | 2 | 300 | 40146360 | ||
| BC144161.1 | - | 2 | 300 | |||

ARSK
Genomic Map