siRNA Details
| Gene Symbol: GPRIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198281.2 | NP_938022.2 | 2 | 2861 | ||
| XM_005262936.3 | XP_005262993.1 | 2 | 3101 | |||
| XM_005262937.4 | XP_005262994.1 | 2 | 3155 | |||
| XM_017008043.1 | XP_016863532.1 | 2 | 4938 | |||
| XM_017008044.1 | XP_016863533.1 | 2 | 2659 | |||
| GenBank mRNA | AB095947.2 | - | 1 | 3426 | ||
| AK124616.1 | - | 2 | 2644 | |||
| BC126371.1 | - | 1 | 2475 | 8992087 | ||

GPRIN3
Genomic Map