siRNA Details
| Gene Symbol: KCTD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198353.2 | NP_938167.1 | 1 | 439 | ||
| XM_011513690.2 | XP_011511992.1 | 1 | 517 | |||
| GenBank mRNA | AK123347.1 | - | 1 | 439 | ||
| AK298831.1 | - | 1 | 125 | |||
| BC132743.1 | - | 1 | 213 | 40146765 | ||
| BC136793.1 | - | 1 | 213 | 9020783 | ||

KCTD8
Genomic Map