siRNA Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270377.1 | NP_001257306.1 | 5 | 2168 | ||
| NM_001270378.1 | NP_001257307.1 | 5 | 2766 | |||
| NM_198401.3 | NP_940683.1 | 6 | 2273 | |||
| XM_006716509.1 | XP_006716572.1 | 5 | 2309 | |||
| XM_017013106.1 | XP_016868595.1 | 5 | 2231 | |||
| GenBank mRNA | AB077205.1 | - | 6 | 2271 | ||
| AK095235.1 | - | 5 | 2143 | |||
| AL832842.1 | - | 5 | 2128 | |||
| BC060843.1 | - | 5 | 2141 | 30341860 | ||
| U79297.1 | - | 1 | 1106 | |||

ANKRD46
Genomic Map