siRNA Details
| Gene Symbol: DGAT2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198512.2 | NP_940914.1 | 7 | 1107 | ||
| GenBank mRNA | AK129500.1 | - | 7 | 1104 | ||
| BG739793.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGAT2L6| Gene Symbol: DGAT2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198512.2 | NP_940914.1 | 7 | 1107 | ||
| GenBank mRNA | AK129500.1 | - | 7 | 1104 | ||
| BG739793.1 | - | 2 | 280 | |||