siRNA Details
| Gene Symbol: CCDC172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198515.2 | NP_940917.1 | 3 | 342 | ||
| XM_006717820.3 | XP_006717883.1 | 3 | 673 | |||
| XM_011539771.2 | XP_011538073.1 | 3 | 670 | |||
| XM_017016206.1 | XP_016871695.1 | 3 | 260 | |||
| GenBank mRNA | AJ457798.1 | - | 2 | 324 | ||
| BC044830.1 | - | 3 | 342 | 5166167 | ||

CCDC172
Genomic Map