siRNA Details
| Gene Symbol: C5orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198566.3 | NP_940968.1 | 3 | 617 | ||
| XM_011514038.2 | XP_011512340.1 | 3 | 526 | |||
| XM_017009445.1 | XP_016864934.1 | 2 | 648 | |||
| GenBank mRNA | AK056925.1 | - | 3 | 532 | ||
| AK303336.1 | - | 2 | 293 | |||
| BC036867.1 | - | 3 | 480 | 5198039 | ||

C5orf34
Genomic Map