siRNA Details
| Gene Symbol: SIMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037298.1 | - | 5 | 1804 | 4823991 | |
| KF151170.1 | - | 3 | 1463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIMC1| Gene Symbol: SIMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037298.1 | - | 5 | 1804 | 4823991 | |
| KF151170.1 | - | 3 | 1463 | |||