siRNA Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308154.1 | NP_001295083.1 | 7 | 1610 | ||
| NM_198686.2 | NP_941959.1 | 7 | 1741 | |||
| XM_017021277.1 | XP_016876766.1 | 8 | 1870 | |||
| GenBank mRNA | BC040679.2 | - | 7 | 1663 | 4817835 | |
| BX248046.1 | - | 6 | 1537 | |||
| BX640825.1 | - | 7 | 1988 | |||

RAB15
Genomic Map