siRNA Details
| Gene Symbol: SLC26A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167962.1 | NP_001161434.1 | 7 | 975 | ||
| NM_001321787.1 | NP_001308716.1 | 7 | 975 | |||
| NM_198999.2 | NP_945350.1 | 7 | 975 | |||
| NM_206883.2 | NP_996766.1 | 7 | 975 | |||
| NM_206884.2 | NP_996767.1 | 7 | 975 | |||
| NM_206885.2 | NP_996768.1 | 7 | 975 | |||
| NR_120441.1 | 6 | 895 | ||||
| NR_120442.1 | 6 | 895 | ||||
| NR_120443.1 | 6 | 895 | ||||
| NR_135801.1 | 7 | 975 | ||||
| NR_135802.1 | 7 | 975 | ||||
| XM_011516170.2 | XP_011514472.1 | 7 | 1765 | |||
| GenBank mRNA | AF523354.1 | - | 7 | 949 | ||
| AK126898.1 | - | 6 | 864 | |||
| AY256823.1 | - | 7 | 949 | |||
| AY256824.1 | - | 7 | 949 | |||
| AY256825.1 | - | 7 | 949 | |||
| AY289133.1 | - | 5 | 721 | |||
| AY289134.1 | - | 5 | 721 | |||
| BC100832.1 | - | 6 | 880 | 40001465 | ||
| BC100833.1 | - | 6 | 880 | 40001467 | ||
| BC100834.1 | - | 6 | 880 | |||
| BC100835.1 | - | 6 | 880 | 40001470 | ||

SLC26A5
Genomic Map