siRNA Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138492.5 | NP_612501.3 | 8 | 1213 | ||
| NM_182960.3 | NP_892005.1 | 8 | 1046 | |||
| NM_205846.2 | NP_995318.1 | 7 | 1010 | |||
| GenBank mRNA | AK095695.1 | - | 8 | 979 | ||

PRELID2
Genomic Map