siRNA Details
| Gene Symbol: ASB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202403.1 | NP_001189332.1 | 5 | 2007 | ||
| NM_017873.3 | NP_060343.1 | 6 | 2094 | |||
| NM_177999.2 | NP_821066.1 | 5 | 1985 | |||
| GenBank mRNA | AK000555.1 | - | 6 | 2091 | ||
| AK225618.1 | - | 5 | 1982 | |||
| AL390135.1 | - | 1 | 367 | |||
| BC001719.1 | - | 5 | 1940 | |||
| BC065913.1 | - | 6 | 1997 | 5211288 | ||
| BX648923.1 | - | 5 | 2615 | |||
| CR627462.1 | - | 1 | 367 | |||

ASB6
Genomic Map