siRNA Details
| Gene Symbol: KRT24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019016.2 | NP_061889.2 | 4 | 992 | ||
| XM_006721739.3 | XP_006721802.1 | 5 | 683 | |||
| XM_011524460.2 | XP_011522762.1 | 5 | 668 | |||
| XM_017024299.1 | XP_016879788.1 | 4 | 977 | |||
| GenBank mRNA | AK000268.1 | - | 4 | 992 | ||
| BC111968.1 | - | 4 | 967 | 8327436 | ||

KRT24
Genomic Map