siRNA Details
| Gene Symbol: ASB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319296.1 | NP_001306225.1 | 5 | 1974 | ||
| NM_001319297.1 | NP_001306226.1 | 4 | 1834 | |||
| NM_001319298.1 | NP_001306227.1 | 5 | 1939 | |||
| NM_001319299.1 | NP_001306228.1 | 5 | 1935 | |||
| NM_001319300.1 | NP_001306229.1 | 5 | 1892 | |||
| NM_001319301.1 | NP_001306230.1 | 5 | 1888 | |||
| NM_024095.4 | NP_077000.1 | 4 | 1838 | |||
| GenBank mRNA | AF464877.1 | - | 4 | 1729 | ||

ASB8
Genomic Map