siRNA Details
| Gene Symbol: CHODL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204174.1 | NP_001191103.1 | 2, 3 | 398 | ||
| NM_001204175.1 | NP_001191104.1 | 2, 3 | 364 | |||
| NM_001204176.1 | NP_001191105.1 | 3, 4 | 464 | |||
| NM_001204177.1 | NP_001191106.1 | 2, 3 | 364 | |||
| NM_001204178.1 | NP_001191107.1 | 3, 4 | 464 | |||
| NM_024944.2 | NP_079220.2 | 2, 3 | 773 | |||
| XM_011529453.2 | XP_011527755.1 | 2, 3 | 1007 | |||
| XM_011529457.2 | XP_011527759.1 | 3, 4 | 770 | |||
| XM_017028273.1 | XP_016883762.1 | 2, 3 | 393 | |||
| GenBank mRNA | AF257472.1 | - | 2, 3 | 773 | ||
| AF523313.1 | - | 3, 4 | 463 | |||
| AF523314.1 | - | 3, 4 | 464 | |||
| AF523315.1 | - | 2, 3 | 365 | |||
| AK022689.1 | - | 3, 4 | 646 | |||
| AK296009.1 | - | 2, 3 | 398 | |||
| AK313725.1 | - | 2, 3 | 421 | |||
| AY358608.1 | - | 2, 3 | 759 | |||
| BC009418.1 | - | 2, 3 | 743 | 3509743 | ||

CHODL
Genomic Map