siRNA Details
| Gene Symbol: C1QTNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031908.4 | NP_114114.2 | 3 | 873 | ||
| XM_011534425.2 | XP_011532727.1 | 4 | 1005 | |||
| XM_011534426.2 | XP_011532728.1 | 5 | 801 | |||
| XM_011534427.2 | XP_011532729.1 | 5 | 896 | |||
| XM_017009013.1 | XP_016864502.1 | 4 | 684 | |||
| GenBank mRNA | AF329836.1 | - | 3 | 881 | ||
| AY358839.1 | - | 3 | 873 | |||
| BC011699.2 | - | 3 | 854 | |||
| BC054506.1 | - | 3 | 847 | |||

C1QTNF2
Genomic Map