siRNA Details
| Gene Symbol: C7orf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026865.1 | 1 | 1924 | |||
| GenBank mRNA | AF063598.1 | - | 1 | 1691 | ||
| AF439977.1 | - | 1 | 1924 | |||
| AK056371.1 | - | 1 | 1840 | |||
| BC031037.1 | - | 1 | 1809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf13| Gene Symbol: C7orf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026865.1 | 1 | 1924 | |||
| GenBank mRNA | AF063598.1 | - | 1 | 1691 | ||
| AF439977.1 | - | 1 | 1924 | |||
| AK056371.1 | - | 1 | 1840 | |||
| BC031037.1 | - | 1 | 1809 | |||