siRNA Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135733.1 | NP_001129205.1 | 5 | 1864 | ||
| NM_033285.3 | NP_150601.1 | 4 | 1826 | |||
| XM_011517386.2 | XP_011515688.1 | 4 | 1678 | |||
| GenBank mRNA | AB017926.1 | - | 3 | 1481 | ||
| AB017927.1 | - | 4 | 1519 | |||
| AF409114.1 | - | 5 | 1836 | |||
| AF409115.1 | - | 4 | 1798 | |||
| AK123608.1 | - | 1 | 18 | |||
| AK125880.1 | - | 5 | 1843 | |||
| AL833652.1 | - | 4 | 1444 | |||

TP53INP1
Genomic Map