siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 2 | 281 | ||
| XM_017010236.1 | XP_016865725.1 | 2 | 472 | |||
| GenBank mRNA | AK124442.1 | - | 2 | 160 | ||
| AK289918.1 | - | 2 | 300 | |||
| AK293853.1 | - | 2 | 235 | |||
| BC069570.1 | - | 2 | 201 | 7262298 | ||
| CD635228.1 | - | 2 | 281 | |||

SLC18B1
Genomic Map