siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 12 | 1385 | ||
| XM_017010236.1 | XP_016865725.1 | 12 | 1576 | |||
| GenBank mRNA | AJ420537.1 | - | 6 | 511 | ||
| AK124442.1 | - | 12 | 1264 | |||
| AK289918.1 | - | 12 | 1404 | |||
| BC069567.1 | - | 8 | 788 | |||
| BC069570.1 | - | 12 | 1305 | 7262298 | ||

SLC18B1
Genomic Map