siRNA Details
| Gene Symbol: CSMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281956.1 | NP_001268885.1 | 4 | 758 | ||
| NM_052896.4 | NP_443128.2 | 4 | 490 | |||
| XM_017000185.1 | XP_016855674.1 | 4 | 752 | |||
| XM_017000186.1 | XP_016855675.1 | 4 | 752 | |||
| XM_017000187.1 | XP_016855676.1 | 4 | 581 | |||
| XM_017000188.1 | XP_016855677.1 | 4 | 490 | |||
| XM_017000189.1 | XP_016855678.1 | 4 | 752 | |||
| XM_017000190.1 | XP_016855679.1 | 4 | 752 | |||
| XM_017000191.1 | XP_016855680.1 | 4 | 752 | |||
| XM_017000192.1 | XP_016855681.1 | 4 | 752 | |||
| XM_017000193.1 | XP_016855682.1 | 4 | 752 | |||
| XM_017000194.1 | XP_016855683.1 | 4 | 490 | |||
| GenBank mRNA | AB212622.1 | - | 4 | 758 | ||
| AY210418.1 | - | 4 | 490 | |||

CSMD2
Genomic Map