siRNA Details
| Gene Symbol: TMEM200A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258276.1 | NP_001245205.1 | 2 | 2803 | ||
| NM_001258277.1 | NP_001245206.1 | 3 | 2867 | |||
| NM_001258278.1 | NP_001245207.1 | 2 | 2822 | |||
| NM_052913.2 | NP_443145.1 | 2 | 4921 | |||
| XM_005266816.2 | XP_005266873.1 | 4 | 2946 | |||
| XM_011535419.2 | XP_011533721.1 | 2 | 2623 | |||
| XM_017010230.1 | XP_016865719.1 | 2 | 3598 | |||
| GenBank mRNA | AB052099.1 | - | 2 | 4921 | ||
| AB067500.1 | - | 3 | 3414 | |||
| BC044246.1 | - | 3 | 2687 | 5760073 | ||

TMEM200A
Genomic Map