siRNA Details
| Gene Symbol: TSGA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304968.1 | NP_001291897.1 | 5 | 1004 | ||
| NM_052933.3 | NP_443165.1 | 4 | 609 | |||
| GenBank mRNA | AF294347.1 | - | 4 | 327 | ||
| AF431815.1 | - | 4 | 609 | |||
| AK093329.1 | - | 5 | 1003 | |||
| BC038415.1 | - | 4 | 609 | 5172129 | ||

TSGA13
Genomic Map