siRNA Details
| Gene Symbol: CYYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320768.1 | NP_001307697.1 | 4 | 1064 | ||
| NM_052954.3 | NP_443186.2 | 4 | 1061 | |||
| NR_135472.1 | 5 | 1231 | ||||
| GenBank mRNA | AF401639.1 | - | 4 | 666 | ||
| AK054581.1 | - | 4 | 941 | |||
| AK223576.1 | - | 4 | 781 | |||
| AK304124.1 | - | 5 | 1111 | |||
| AL833200.1 | - | 4 | 872 | |||
| AY061853.1 | - | 4 | 606 | |||
| BC036761.1 | - | 4 | 849 | 5588436 | ||

CYYR1
Genomic Map