siRNA Details
| Gene Symbol: ACSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318890.1 | NP_001305819.1 | 6 | 1020 | ||
| NM_052956.2 | NP_443188.2 | 5 | 884 | |||
| NR_134918.1 | 6 | 945 | ||||
| XM_006721016.2 | XP_006721079.1 | 5 | 863 | |||
| XM_006721017.2 | XP_006721080.1 | 5 | 863 | |||
| XM_006721018.2 | XP_006721081.1 | 3 | 242 | |||
| XM_011545729.2 | XP_011544031.1 | 5 | 863 | |||
| XM_011545730.2 | XP_011544032.1 | 5 | 863 | |||
| XM_011545731.2 | XP_011544033.1 | 3 | 1059 | |||
| XM_011545732.2 | XP_011544034.1 | 4 | 301 | |||
| XM_017022914.1 | XP_016878403.1 | 5 | 863 | |||
| XM_017022915.1 | XP_016878404.1 | 4 | 1711 | |||
| GenBank mRNA | AB059429.1 | - | 5 | 884 | ||
| AK092574.1 | - | 3 | 242 | |||
| AK314282.1 | - | 6 | 976 | |||
| BC125177.1 | - | 6 | 945 | |||
| BC125178.1 | - | 5 | 884 | 40121956 | ||

ACSM1
Genomic Map