siRNA Details
| Gene Symbol: ACRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052957.4 | NP_443189.1 | 8 | 864 | ||
| XM_006724716.3 | XP_006724779.1 | 8 | 417 | |||
| XM_011531063.2 | XP_011529365.1 | 7 | 684 | |||
| XM_011531064.1 | XP_011529366.1 | 8 | 467 | |||
| XM_011531066.2 | XP_011529368.1 | 5 | 603 | |||
| XM_011531067.2 | XP_011529369.1 | 5 | 603 | |||
| XM_017029965.1 | XP_016885454.1 | 9 | 518 | |||
| GenBank mRNA | AJ311392.1 | - | 7 | 365 | ||
| AK097433.1 | - | 8 | 478 | |||
| AK127607.1 | - | 7 | 900 | |||
| BC136259.1 | - | 8 | 432 | 9020246 | ||

ACRC
Genomic Map