siRNA Details
| Gene Symbol: FAM129A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052966.3 | NP_443198.1 | 14 | 3763 | ||
| XM_011509140.2 | XP_011507442.1 | 15 | 4638 | |||
| XM_011509141.2 | XP_011507443.1 | 14 | 4527 | |||
| GenBank mRNA | AB050477.1 | - | 14 | 3689 | ||
| AF288391.1 | - | 14 | 3727 | |||
| AK022527.1 | - | 14 | 3601 | |||
| AK074139.1 | - | 2 | 1971 | |||
| AK095547.1 | - | 10 | 2052 | |||
| AL137572.1 | - | 1 | 387 | |||
| BC030531.1 | - | 14 | 3619 | 5212338 | ||

FAM129A
Genomic Map