siRNA Details
| Gene Symbol: HSPA12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197327.1 | NP_001184256.1 | 13 | 2441 | ||
| NM_001318322.1 | NP_001305251.1 | 12 | 2346 | |||
| NM_052970.4 | NP_443202.3 | 13 | 2444 | |||
| XM_011529151.2 | XP_011527453.1 | 10 | 2142 | |||
| XM_017027632.1 | XP_016883121.1 | 13 | 2359 | |||
| GenBank mRNA | AK056712.1 | - | 13 | 2381 | ||
| BC090857.1 | - | 12 | 2305 | |||
| BC110881.1 | - | 13 | 2393 | 5725594 | ||
| BC117284.1 | - | 13 | 2342 | 40125835 | ||
| BC126263.1 | - | 13 | 2342 | 8991979 | ||
| BC143932.1 | - | 13 | 2339 | 9052450 | ||

HSPA12B
Genomic Map