siRNA Details
| Gene Symbol: ODF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286136.1 | NP_001273065.1 | 6 | 1041 | ||
| NM_053280.4 | NP_444510.2 | 7 | 1182 | |||
| GenBank mRNA | AB067774.1 | - | 7 | 1289 | ||
| AL133658.1 | - | 3 | 2950 | |||
| AY713299.1 | - | 7 | 1157 | |||

ODF3
Genomic Map