siRNA Details
| Gene Symbol: ODF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286136.1 | NP_001273065.1 | 2, 3 | 402 | ||
| NM_053280.4 | NP_444510.2 | 2, 3 | 402 | |||
| GenBank mRNA | AB067774.1 | - | 2, 3 | 509 | ||
| AL133658.1 | - | 1, 2 | 621 | |||
| AY713299.1 | - | 2, 3 | 377 | |||
| BC126222.1 | - | 2, 3 | 210 | 8991938 | ||
| BC126248.1 | - | 2, 3 | 210 | 8991964 | ||
| BC144022.1 | - | 2, 3 | 210 | 9052542 | ||
| FJ436414.1 | - | 1, 2 | 110 | |||

ODF3
Genomic Map