siRNA Details
| Gene Symbol: DACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139515.1 | NP_001132987.1 | 11, 12 | 1314 | ||
| NM_053281.3 | NP_444511.1 | 11, 12 | 1928 | |||
| XM_011530846.2 | XP_011529148.1 | 11, 12 | 1957 | |||
| XM_011530847.2 | XP_011529149.1 | 12 | 6297 | |||
| XM_011530848.2 | XP_011529150.1 | 12 | 6267 | |||
| XM_017029254.1 | XP_016884743.1 | 10, 11 | 1918 | |||
| XM_017029255.1 | XP_016884744.1 | 10, 11 | 1888 | |||
| XM_017029256.1 | XP_016884745.1 | 11 | 6228 | |||
| GenBank mRNA | AF428101.1 | - | 11, 12 | 1758 | ||
| AK055953.1 | - | 10, 11 | 1275 | |||
| AK290596.1 | - | 11, 12 | 1928 | |||
| AK298787.1 | - | 11, 12 | 1314 | |||
| AL834456.1 | - | 4, 5 | 520 | |||
| BC114950.1 | - | 11, 12 | 1837 | 40006598 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001289732, NM_033605, NM_001289734, AK042401, AF257217, and others.

DACH2
Genomic Map