siRNA Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142883.1 | NP_001136355.1 | 5, 6 | 1126 | ||
| NM_054111.4 | NP_473452.2 | 4, 5 | 997 | |||
| XM_005248842.3 | XP_005248899.1 | 5, 6 | 906 | |||
| XM_005248843.3 | XP_005248900.1 | 3, 4 | 1017 | |||
| XM_011514295.2 | XP_011512597.1 | 4, 5 | 1009 | |||
| GenBank mRNA | AF393812.1 | - | 3, 4 | 692 | ||
| AK056586.2 | - | 4, 5 | 916 | |||
| AK223572.1 | - | 4, 5 | 920 | |||
| AK289935.1 | - | 5, 6 | 1046 | |||
| BC089389.1 | - | 4, 5 | 997 | 30530548 | ||

IP6K3
Genomic Map