siRNA Details
| Gene Symbol: KLHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057161.3 | NP_476502.1 | 3 | 521 | ||
| NR_040101.1 | 3 | 521 | ||||
| GenBank mRNA | AB055925.1 | - | 3 | 385 | ||
| AK290384.1 | - | 3 | 377 | |||
| AK302336.1 | - | 2 | 160 | |||
| AK308917.1 | - | 3 | 407 | |||
| BC000295.2 | - | 3 | 341 | 2819570 | ||
| BC001789.1 | - | 3 | 356 | 3355470 | ||
| BC001793.2 | - | 3 | 319 | 3352397 | ||
| BC007296.1 | - | 3 | 371 | 3349731 | ||
| BC009460.1 | - | 3 | 369 | 3537223 | ||
| BC021546.1 | - | 3 | 358 | 3351635 | ||
| BC041793.1 | - | 3 | 375 | 5288540 | ||
| BC045612.1 | - | 3 | 375 | 5313255 | ||
| DB057971.1 | - | 3 | 394 | |||
| DC395338.1 | - | 3 | 521 | |||

KLHDC3
Genomic Map