siRNA Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080476.4 | NP_536724.1 | 8 | 915 | ||
| XM_011528542.2 | XP_011526844.1 | 2 | 206 | |||
| XM_017027664.1 | XP_016883153.1 | 8 | 733 | |||
| GenBank mRNA | AB086842.1 | - | 8 | 721 | ||
| AK075507.1 | - | 8 | 740 | |||
| AK301900.1 | - | 8 | 715 | |||
| AK308627.1 | - | 8 | 780 | |||
| AY339061.1 | - | 7 | 655 | |||
| AY358816.1 | - | 8 | 745 | |||
| AY422169.1 | - | 8 | 915 | |||
| BC030512.1 | - | 8 | 726 | 4942673 | ||

PIGU
Genomic Map