siRNA Details
| Gene Symbol: CST9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080610.2 | NP_542177.1 | 1 | 406 | ||
| GenBank mRNA | AK312112.1 | - | 1 | 404 | ||
| AY358615.1 | - | 1 | 349 | |||
| BC029656.1 | - | 1 | 406 | 5163974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST9L| Gene Symbol: CST9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080610.2 | NP_542177.1 | 1 | 406 | ||
| GenBank mRNA | AK312112.1 | - | 1 | 404 | ||
| AY358615.1 | - | 1 | 349 | |||
| BC029656.1 | - | 1 | 406 | 5163974 | ||