siRNA Details
| Gene Symbol: GAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081573.2 | NP_001075042.1 | 10 | 4110 | ||
| NM_001282283.1 | NP_001269212.1 | 9 | 3993 | |||
| NM_080612.3 | NP_542179.1 | 10 | 4107 | |||
| NR_104114.1 | 9 | 3599 | ||||
| XM_006724804.1 | XP_006724867.1 | 11 | 2821 | |||
| XM_011531103.1 | XP_011529405.1 | 11 | 3962 | |||
| XM_011531106.1 | XP_011529408.1 | 10 | 3845 | |||
| XM_017029276.1 | XP_016884765.1 | 11 | 2812 | |||
| GenBank mRNA | AL832051.1 | - | 9 | 3560 | ||
| AY057989.1 | - | 10 | 4081 | |||

GAB3
Genomic Map