siRNA Details
| Gene Symbol: NOL4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256798.1 | NP_001243727.1 | 11 | 2525 | ||
| NM_080616.4 | NP_542183.2 | 8 | 1864 | |||
| GenBank mRNA | AK056286.1 | - | 8 | 1669 | ||
| AL122043.2 | - | 1 | 130 | |||
| BC065370.1 | - | 8 | 1221 | 6261963 | ||

NOL4L
Genomic Map