siRNA Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080626.5 | NP_542193.3 | 3 | 1099 | ||
| GenBank mRNA | AF284094.1 | - | 3 | 1043 | ||
| AK055703.1 | - | 1 | 620 | |||
| BC034525.1 | - | 3 | 1076 | 4778946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRI3BP| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080626.5 | NP_542193.3 | 3 | 1099 | ||
| GenBank mRNA | AF284094.1 | - | 3 | 1043 | ||
| AK055703.1 | - | 1 | 620 | |||
| BC034525.1 | - | 3 | 1076 | 4778946 | ||