siRNA Details
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080725.2 | NP_542763.1 | 2 | 837 | ||
| GenBank mRNA | AK098418.1 | - | 1 | 38 | ||
| AK127435.1 | - | 2 | 682 | |||
| AL833944.1 | - | 2 | 713 | |||
| AY427953.1 | - | 1 | 38 | |||
| BC017001.1 | - | 2 | 632 | |||
| BC032604.1 | - | 2 | 668 | 5535883 | ||
| BC047707.1 | - | 2 | 657 | 5735551 | ||

SRXN1
Genomic Map