siRNA Details
| Gene Symbol: TMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080751.2 | NP_542789.2 | 11, 12 | 1429 | ||
| XM_005260660.3 | XP_005260717.1 | 9, 10 | 1623 | |||
| GenBank mRNA | AF417580.2 | - | 11, 12 | 1429 | ||
| AK094789.1 | - | 1, 2 | 137 | |||
| AK294013.1 | - | 9, 10 | 1175 | |||
| AK316261.1 | - | 9, 10 | 1145 | |||

TMC2
Genomic Map