siRNA Details
| Gene Symbol: ZSWIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080752.3 | NP_542790.2 | 2 | 501 | ||
| NR_037628.1 | 3 | 656 | ||||
| GenBank mRNA | AK056641.1 | - | 2 | 466 | ||
| AK302767.1 | - | 3 | 620 | |||
| AK316214.1 | - | 3 | 619 | |||
| BC027611.2 | - | 2 | 458 | 4829980 | ||
| BX385024.2 | - | 2 | 459 | |||
| DA520427.1 | - | 2 | 501 | |||

ZSWIM3
Genomic Map