siRNA Details
| Gene Symbol: DPCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080870.3 | NP_543146.2 | 3 | 5038 | ||
| GenBank mRNA | AB064272.1 | - | 2 | 1566 | ||
| AL833738.1 | - | 2 | 2380 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPCR1| Gene Symbol: DPCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080870.3 | NP_543146.2 | 3 | 5038 | ||
| GenBank mRNA | AB064272.1 | - | 2 | 1566 | ||
| AL833738.1 | - | 2 | 2380 | |||