siRNA Details
| Gene Symbol: STATH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009181.1 | NP_001009181.1 | 5 | 448 | ||
| NM_003154.2 | NP_003145.1 | 6 | 478 | |||
| GenBank mRNA | BC067219.1 | - | 6 | 444 | 30328926 | |
| BX649104.1 | - | 2 | 4445 | |||
| CB958641.1 | - | 6 | 445 | |||
| CB986973.1 | - | 482 | ||||
| M18078.1 | - | 6 | 440 | |||
| M18371.1 | - | 6 | 424 | |||

STATH
Genomic Map