siRNA Details
| Gene Symbol: CD109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159587.2 | NP_001153059.1 | 4 | 550 | ||
| NM_133493.4 | NP_598000.2 | 4 | 550 | |||
| XM_005248659.3 | XP_005248716.1 | 5 | 697 | |||
| XM_011535473.1 | XP_011533775.1 | 4 | 395 | |||
| XM_017010283.1 | XP_016865772.1 | 2 | 162 | |||
| GenBank mRNA | AF410459.1 | - | 4 | 531 | ||
| AK095888.1 | - | 4 | 484 | |||
| AK313636.1 | - | 4 | 530 | |||
| AY149920.1 | - | 4 | 419 | |||
| AY788891.1 | - | 4 | 444 | |||
| EF553520.1 | - | 4 | 449 | |||

CD109
Genomic Map