siRNA Details
| Gene Symbol: NEK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133494.2 | NP_598001.1 | 10 | 1237 | ||
| XM_011509209.1 | XP_011507511.1 | 6 | 789 | |||
| XM_017000344.1 | XP_016855833.1 | 10 | 1239 | |||
| XM_017000345.1 | XP_016855834.1 | 11 | 1346 | |||
| XM_017000346.1 | XP_016855835.1 | 9 | 1125 | |||
| GenBank mRNA | AB062450.1 | - | 9 | 956 | ||

NEK7
Genomic Map